Double bossheads
With nickel plated brass T-screws, Aluminium alloy, powder coated, 13,0 mm
Catalog # 241-7203
Supplier: USBECK Laborgeräte
Jump to:
Product Details & Documents
For stand assembly.
- Quick, easy assembly and disassembly
- Robust, durable
- Wide range of different materials available for many applications
Specifications
- Description:With nickel plated brass T-screws
- Material:Aluminium alloy, powder coated
- Seq:5
- For rod Ø:13,0 mm