Order Entry
Export
Orders LinkContactUsLinkComponent
Burner, Teclu

Burner, Teclu

Teclu burner with air regulation needle valve, all gases, tubing Ø 14 mm, All gases, 17 mm
Catalog # 451-0088
Supplier:   USBECK Laborgeräte
Burner, Teclu
Catalog # 451-0088
Burner, Teclu
Burner, Teclu

Teclu burner with air regulation needle valve, all gases, tubing Ø 14 mm, All gases, 17 mm

Price
55,70 €
Log in for availability
1 * 1 items
Product Details & Documents

Nickel plated brass base, in powder coated grey.

  • With air regulation valve and throttle screw
  • Without pilot light

Height: 165 mm

Specifications
  • Description:
    Teclu burner with air regulation needle valve, all gases, tubing Ø 14 mm
  • Head Ø:
    17 mm
  • For:
    All gases