Orders LinkContactUsLinkComponent
Comparator kits, Lovibond® system 2000
Comparator kits, Lovibond® system 2000
Platinum-Cobalt Units (Low Range, 0 - 70 mg Pt/l)) Nessleriser 2250 with Daylight 2000 Lighting unit and 250 mm Nessler cylinders; Pt-Co discs CAA (0 - 30 mg Pt/l), CAB (30 - 70 mg Pt/l); stand for using Nessleriser with natural lighting, AF328
Catalog # 633-0022
Supplier:   Lovibond Tintometer
Comparator kits, Lovibond® system 2000
Catalog # 633-0022
Comparator kits, Lovibond® system 2000
Comparator kits, Lovibond® system 2000
Platinum-Cobalt Units (Low Range, 0 - 70 mg Pt/l)) Nessleriser 2250 with Daylight 2000 Lighting unit and 250 mm Nessler cylinders; Pt-Co discs CAA (0 - 30 mg Pt/l), CAB (30 - 70 mg Pt/l); stand for using Nessleriser with natural lighting, AF328
Price
2 480,00 €
Log in for availability
1 * 1 items
Product Details & Documents
Available for many of the commonly used colour scales, these kits are a convenient means of ordering the complete range of equipment required. Each system includes.
  • A visual comparator instrument which brings the sample and colour standards into the same field of view for more accurate colour matching
  • A selection of colour standards - stable-coloured glass filters in test discs, which have been pre-calibrated to give direct readings over set measuring ranges
  • A standardised benchtop light source to ensure constant, uniform lighting conditions for colour matching and, therefore, consistent and accurate readings
  • Glassware of the appropriate type and path length
Specifications
  • Description:
    Platinum-Cobalt Units (Low Range, 0 - 70 mg Pt/l)) Nessleriser 2250 with Daylight 2000 Lighting unit and 250 mm Nessler cylinders; Pt-Co discs CAA (0 - 30 mg Pt/l), CAB (30 - 70 mg Pt/l); stand for using Nessleriser with natural lighting
  • Type:
    AF328