Skip to Header
Skip to Main Content
Skip to Footer
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
Orders Link
ContactUsLinkComponent
Home
Media for Microbiology
Additives and Supplements for Microbiology
Blood
Blood
Print
Blood
Horse Serum, 100 mL
Catalog # OXOISR0035C
Supplier:
THERMO FISHER DIAGNOSTICS
Blood
Catalog # OXOISR0035C
Supplier:
THERMO FISHER DIAGNOSTICS
CAS Number:
Expand image
Return to Top
Blood
Horse Serum, 100 mL
Specifications
Add to cart
Jump to:
Specifications
CAS Number:
Price
91,70 €
Log in
for availability
Unit of Measure
1 * 100 mL
-
+
Add to cart
Other Product Family Options Available
Return Policy
Product Details & Documents
Documents
Certificates
Search by Lot Number
Datasheet
OXOISR0048C_TDS_EN
Specifications
Specifications
Pk:
100 mL
Description:
Horse Serum
Specifications