0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
Orders Link
ContactUsLinkComponent
Labels
Hazardous substance labels
Hazardous substance labels
Hazard labels, Paper, Red/yellow, 76×25 mm
TIMERA-1
:
PRECISION DYNAMICS TIMEM
Hazardous substance labels
TIMERA-1
:
PRECISION DYNAMICS TIMEM
Hazardous substance labels
Hazard labels, Paper, Red/yellow, 76×25 mm
49,00 €
1 * 1 Roll
-
+
Colour:
Red/yellow
Description:
Hazard labels
Dimensions:
76×25 mm
Material:
Paper