Order Entry
Export
Orders LinkContactUsLinkComponent
Precision probes for 735

Precision probes for 735

PT100 glass-coated laboratory probe with replaceable glass pipe, −50…+400 °C, ±0,2 °C
±0,2% mv (+200...+800 °C), 6×200 mm
Catalog # 620-3325
Supplier:   Testo
Precision probes for 735
Catalog # 620-3325
Supplier:  Testo
Precision probes for 735
Precision probes for 735

PT100 glass-coated laboratory probe with replaceable glass pipe, −50…+400 °C, ±0,2 °C
±0,2% mv (+200...+800 °C), 6×200 mm

Price
134,00 €
Log in for availability
1 * 1 items
Specifications
  • Range:
    −50…+400 °C
  • Description:
    PT100 glass-coated laboratory probe with replaceable glass pipe
  • Accuracy:
    ±0,2 °C
    ±0,2% mv (+200...+800 °C)
  • Ø×L:
    6×200 mm