Order Entry
Export
Orders LinkContactUsLinkComponent
Precision probes for 735

Precision probes for 735

TC type T flexible air probe with PTFE cable, −50…+250 °C, ±0,3 °C (−20...+70 °C)
±(0,7 °C (±0,5% mv), 1,5×2000 mm
Catalog # 628-3299
Supplier:   Testo
Precision probes for 735
Catalog # 628-3299
Supplier:  Testo
Precision probes for 735
Precision probes for 735

TC type T flexible air probe with PTFE cable, −50…+250 °C, ±0,3 °C (−20...+70 °C)
±(0,7 °C (±0,5% mv), 1,5×2000 mm

Price
50,00 €
Log in for availability
1 * 1 items
Specifications
  • Range:
    −50…+250 °C
  • Description:
    TC type T flexible air probe with PTFE cable
  • Seq:
    01
  • Accuracy:
    ±0,3 °C (−20...+70 °C)
    ±(0,7 °C (±0,5% mv)
  • Ø×L:
    1,5×2000 mm