Order Entry
Export
Orders LinkContactUsLinkComponent
Probes for 845

Probes for 845

K-type fibreglass, flexible thermocouple with TC adapter for surface, −50…+400 °C, −40...+1200 °C, 1,5×800 mm
Catalog # 620-3425
Supplier:   Testo
Probes for 845
Catalog # 620-3425
Supplier:  Testo
Probes for 845
Probes for 845

K-type fibreglass, flexible thermocouple with TC adapter for surface, −50…+400 °C, −40...+1200 °C, 1,5×800 mm

Price
22,00 €
Log in for availability
1 * 1 items
Specifications
  • Range:
    −50…+400 °C
  • Description:
    K-type fibreglass, flexible thermocouple with TC adapter for surface
  • Seq:
    23
  • Accuracy:
    −40...+1200 °C
  • Application:
    Surface
  • Ø×L:
    1,5×800 mm