Cryo-Gloves® Cold Resistant Gloves
These hand-specific gloves protect hands and arms against the hazards of working in and with ultra-cold temperatures down to –196 °C. Lightweight, breathable, and comfortable, they provide protection from liquid splashes and cryogenic temperatures but are not safe for immersion in cryogenic liquids.
- Provide warmth while retaining flexibility even after prolonged exposure to cryogenic atmospheres
- Cryo-Gloves® allow sufficient dexterity to handle small objects and controls
- Inner cuffs (except for the wrist length models) contain a single layer of insulation material, a scrim backing and a nylon taffeta lining
- Wrist length models feature a knitted cuff, all other models feature a gauntlet cuff
- Sewn construction
Cryo-Gloves® are offered in three configurations:
Water resistant: These gloves comprise of an outer shell consisting of nylon Taslan® with a liquid-proof, breathable membrane on the inner surface, and an inner insulated glove consisting of three layers (non woven synthetic insulation material + lining material).
Waterproof: These gloves have the same construction as the water-resistant gloves but additionally feature a seamless, breathable, waterproof lining.
Industrial: A rugged, abrasion resistant outer shell and a thin, seamless 100% waterproof liner provide extra protection from cryogenic splashes and an excellent grip for heavy-duty applications.
EN 388, EN 511, Cat. III
: EN 388, EN 511, Cat. III
- Glove size:XL
- Colour:Blue
- Length:345 - 390 mm
- Seq:25
- Model:Mid-arm
- Cat. No.:112-2711
- Disposable / reusable:Reusable
- EN norm:EN 388 (1323), EN 511 (321)
- Ambidextrous / hand-specific:Hand-specific



