Orders LinkContactUsLinkComponent
Plates, Nunc-Immuno™ TSP (transferable solid phases)
Plates, Nunc-Immuno™ TSP (transferable solid phases)
Nunc-Immuno™ TSP plates, 96-well, with MaxiSorp™ surface
Catalog # 735-0074
Plates, Nunc-Immuno™ TSP (transferable solid phases)
Catalog # 735-0074
Plates, Nunc-Immuno™ TSP (transferable solid phases)
Plates, Nunc-Immuno™ TSP (transferable solid phases)
Nunc-Immuno™ TSP plates, 96-well, with MaxiSorp™ surface
Price
1 440,00 €
Log in for availability
1 * 210 items
Product Details & Documents
PS. For simultaneous testing of analytes using ELISA technique. Also available sterile for screening of hybridoma cultures for antibody production.
  • Uniform adsorption properties
  • Plates have 96 round bottom wells with total surface area of 95 mm²
  • Sterile plates supplied mounted in the tray
MaxiSorp™ surface has a high affinity to molecules with mixed hydrophilic/hydrophobic domains.
Colour: Clear
Colour: Clear
Specifications
  • Description:
    Nunc-Immuno™ TSP plates, 96-well, with MaxiSorp™ surface