Order Entry
Export
Orders LinkContactUsLinkComponent
Tubes, Nunc-Immuno™, Nunc™

Tubes, Nunc-Immuno™, Nunc™

5,0 ml, Nunc StarTubes™, MaxiSorp™, 12×75 mm
Catalog # 391-8281
   Low Manufacturing Impact
Tubes, Nunc-Immuno™, Nunc™
Catalog # 391-8281
   Low Manufacturing Impact
Tubes, Nunc-Immuno™, Nunc™
Tubes, Nunc-Immuno™, Nunc™

5,0 ml, Nunc StarTubes™, MaxiSorp™, 12×75 mm

Price
344,00 €
Log in for availability
1 * 3.000 items
Product Details & Documents

PS with a choice of MaxiSorp™ (hydrophilic) or PolySorp™ (hydrophobic) surfaces.

  • Nunc StarTubes™ with an increased internal surface, give higher sensitivity and faster assay times
  • Very good optical quality and MaxiSorp™ versions have certified binding homogeneity
  • Compatible with standard equipment

Nunc-Immuno™ tubes are used for solid phase immunoassay methods such as IRMA, ELISA and ILMA methods.

Specifications
  • Capacity:
    5,0 ml
  • Description:
    Nunc StarTubes™, MaxiSorp™
  • Seq:
    03
  • Ø ext.×H:
    12×75 mm
  • Cat. No.:
    391-8281
  • Environmental certification:
    ISO 14001 (Certificate Registration No.CERT-IT000330)
  • Environmentally Preferable:
    Low Manufacturing Impact
Accessories

Product Information

  • Catalog #
    Availability
    Price
  • Stoppers, PE, for tubes 15 mm external diameter
    Stoppers, PE, for tubes 15 mm external diameter
    391-8062
    Log in for availability
    1 * 2.400 items - 194,00 €

Product Information

  • Catalog #
    Availability
    Price
  • Stoppers, PE, for tubes 12 mm external diameter
    Stoppers, PE, for tubes 12 mm external diameter
    732-2680
    Log in for availability
    1 * 3.000 items - 186,00 €