Tubes, low binding, Nunc-Immuno™, Nunc™
12 ml, MiniSorp™, 15×100 mm
Catalog # 735-0047
Supplier: Thermo Fisher Scientific
Tubes, low binding, Nunc-Immuno™, Nunc™
Catalog # 735-0047
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Graduated polyethylene (PE).
- Surface has lower affinity to proteins than glass and other plastics
- No blocking required
- Compatible with standard equipment
Nunc-Immuno™ MiniSorp™ tubes are suitable for liquid phase immuno-assay methods including RIA.
Specifications
- Capacity:12 ml
- Description:MiniSorp™
- Seq:03
- Ø ext.×H:15×100 mm
- Cat. No.:735-0047
- Environmental certification:ISO 14001 (Certificate Registration No.CERT-IT000330)
- Environmentally Preferable:Low Manufacturing Impact
Accessories
Product Information
- Catalog #AvailabilityPrice
- Stoppers, PE, for tubes 15 mm external diameterStoppers, PE, for tubes 15 mm external diameter391-8062Log in for availability1 * 2.400 items - 194,00 €
Product Information
- Catalog #AvailabilityPrice
- Stoppers, PE, for tubes 12 mm external diameterStoppers, PE, for tubes 12 mm external diameter732-2680Log in for availability1 * 3.000 items - 186,00 €
Product Information
- Catalog #AvailabilityPrice
- Stoppers, PE, for tubes 11 mm external diameterStoppers, PE, for tubes 11 mm external diameter391-8056Log in for availability1 * 3.600 items - 234,00 €



