Tubes, Nunc-Immuno™, Nunc™
5,0 ml, Tubes MaxiSorp™, 12×75 mm
Supplier: Thermo Fisher Scientific
Tubes, Nunc-Immuno™, Nunc™
Catalog # 735-0018
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
PS with a choice of MaxiSorp™ (hydrophilic) or PolySorp™ (hydrophobic) surfaces.
- Nunc StarTubes™ with an increased internal surface, give higher sensitivity and faster assay times
- Very good optical quality and MaxiSorp™ versions have certified binding homogeneity
- Compatible with standard equipment
Nunc-Immuno™ tubes are used for solid phase immunoassay methods such as IRMA, ELISA and ILMA methods.
Specifications
- Capacity:5,0 ml
- Description:Tubes MaxiSorp™
- Seq:02
- Ø ext.×H:12×75 mm
- Cat. No.:735-0018
- Environmental certification:ISO 14001 (Certificate Registration No.CERT-IT000330)
- Environmentally Preferable:Low Manufacturing Impact
Accessories
Product Information
- Catalog #AvailabilityPrice
- Stoppers, PE, for tubes 15 mm external diameterStoppers, PE, for tubes 15 mm external diameter391-8062Log in for availability1 * 2.400 items - 194,00 €
Product Information
- Catalog #AvailabilityPrice
- Stoppers, PE, for tubes 12 mm external diameterStoppers, PE, for tubes 12 mm external diameter732-2680Log in for availability1 * 3.000 items - 186,00 €



