Tubes, low binding, Nunc-Immuno™, Nunc™
5 ml, MiniSorp™, 12×75 mm
735-0017
Tubes, low binding, Nunc-Immuno™, Nunc™
735-0017
Graduated polyethylene (PE).
- Surface has lower affinity to proteins than glass and other plastics
- No blocking required
- Compatible with standard equipment
Nunc-Immuno™ MiniSorp™ tubes are suitable for liquid phase immuno-assay methods including RIA.
- Capacity:5 ml
- Description:MiniSorp™
- Seq:02
- Ø ext.×H:12×75 mm
- Cat. No.:735-0017
- Environmental certification:ISO 14001 (Certificate Registration No.CERT-IT000330)
- Environmentally Preferable:Low Manufacturing Impact



