Lids for Nunc™ microplates
Lid, PS, with cut-off corners and evaporation barrier, for use with 384 MicroWell™ plates, Clear, 7,9 mm, Sterile
Catalog # 732-2732
Supplier: Thermo Fisher Scientific
Lids for Nunc™ microplates
Catalog # 732-2732
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Documents
Specifications
- Sterility:Sterile
- Colour:Clear
- Description:Lid, PS, with cut-off corners and evaporation barrier, for use with 384 MicroWell™ plates
- Height:7,9 mm
- Seq:07
- Cat. No.:732-2732
- Environmental certification:NAR Certificate of Retirement
- Environmentally Preferable:Low Manufacturing Impact