Order Entry
Export
Orders LinkContactUsLinkComponent
Storage vials, Nunc™

Storage vials, Nunc™

2,00 ml, 13×49 mm, Yes
Catalog # 391-7589
   Low Manufacturing Impact
Storage vials, Nunc™
Catalog # 391-7589
   Low Manufacturing Impact
Storage vials, Nunc™
Storage vials, Nunc™

2,00 ml, 13×49 mm, Yes

Price
482,00 €
Log in for availability
1 * 500 items
Product Details & Documents

High purity PPCO (PP copolymer) vial, PE screw cap. Excellent for centrifugation and storage of PCR reagents, restriction enzymes and other diagnostic and biochemical reagents or samples. Available in 0,5 and 2,0 ml sizes with frosted writing area; furthermore 2,0 ml size have graduations at 0,5 and 1,0 ml.

  • Leakproof screw closure even without an O-ring
  • One full turn removes the closure, giving easy access to contents

Can be centrifuged to a max. of 13000×g.

Can be used from −40 to +95 °C.

Can be used from -40 to +95 °C.

Specifications
  • Cap included:
    Yes
  • Ø×H:
    13×49 mm
  • Capacity:
    2,00 ml
  • Cat. No.:
    391-7589
  • Environmental certification:
    NAR Certificate of Retirement
  • Environmentally Preferable:
    Low Manufacturing Impact