Storage vials, Nunc™
0,50 ml, 13×49 mm, Yes
Catalog # 391-8348
Jump to:
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
VIAL 0,5ML CRYOBANK 2D CODED RACKED STERILE 1 * 960 items Catalog #479-0588
Product 479-0588 is similar to 391-8348 but manufacturer and specifications may vary.
Screw cap microTube centrifuge 0.5 ml 20,000g free-standing clear non-steril no cap 1 * 500 items Catalog #525-0643
Product 525-0643 is similar to 391-8348 but manufacturer and specifications may vary.
Screw cap microTube centrifuge 0.5 ml 20,000g conical-bottom clear steril attached cap 1 * 500 items Catalog #525-0642
Product 525-0642 is similar to 391-8348 but manufacturer and specifications may vary.
Product Details & Documents
High purity PPCO (PP copolymer) vial, PE screw cap. Excellent for centrifugation and storage of PCR reagents, restriction enzymes and other diagnostic and biochemical reagents or samples. Available in 0,5 and 2,0 ml sizes with frosted writing area; furthermore 2,0 ml size have graduations at 0,5 and 1,0 ml.
- Leakproof screw closure even without an O-ring
- One full turn removes the closure, giving easy access to contents
Can be centrifuged to a max. of 13000×g.
Can be used from −40 to +95 °C.
Can be used from -40 to +95 °C.
Specifications
- Cap included:Yes
- Ø×H:13×49 mm
- Capacity:0,50 ml



