Lids for Nunc™ microplates
Lid, PS, with cut-off corners, condensation rings and evaporation barrier, for use with 96 MicroWell™ plates, Clear, 7,9 mm, Non sterile
Supplier: Thermo Fisher Scientific
Lids for Nunc™ microplates
Catalog # 734-2184
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Documents
Specifications
- Sterility:Non sterile
- Colour:Clear
- Description:Lid, PS, with cut-off corners, condensation rings and evaporation barrier, for use with 96 MicroWell™ plates
- Height:7,9 mm
- Seq:04
- Cat. No.:734-2184
- Environmental certification:ISO 14001 (Certificate Registration No.CERT-IT000330)
- Environmentally Preferable:Low Manufacturing Impact