Orders LinkContactUsLinkComponent
Inoculating needles and loops, Nunc™
Inoculating needles and loops, Nunc™
Loops, 1 µl, Transparent, 12/sleeve
Catalog # 710-2508
   Low Manufacturing Impact
Inoculating needles and loops, Nunc™
Catalog # 710-2508
   Low Manufacturing Impact
Inoculating needles and loops, Nunc™
Inoculating needles and loops, Nunc™
Loops, 1 µl, Transparent, 12/sleeve
Price
472,00 €
Log in for availability
1 * 2.400 items
Product Details & Documents
PS, sterile. Disposable, problem free inoculation with smooth loop surface.
  • Meet semi-quantitative standard for handling samples
  • Surface treated for enhanced droplet adhesion
  • Non toxic
Specifications
  • Packed:
    12/sleeve
  • Capacity:
    1 µl
  • Colour:
    Transparent
  • Description:
    Loops
  • Seq:
    01
  • Cat. No.:
    710-2508
  • Environmental certification:
    ISO 14001 (Certificate Registration No.CERT-IT000330)
  • Environmentally Preferable:
    Low Manufacturing Impact