Orders LinkContactUsLinkComponent
Inoculating needles and loops, Nunc™
Inoculating needles and loops, Nunc™
Needles, -, Yellow, 50/sleeve
Catalog # 734-2139
   Low Manufacturing Impact
Inoculating needles and loops, Nunc™
Catalog # 734-2139
   Low Manufacturing Impact
Inoculating needles and loops, Nunc™
Inoculating needles and loops, Nunc™
Needles, -, Yellow, 50/sleeve
Price
527,00 €
Log in for availability
1 * 4.000 items
Product Details & Documents
PS, sterile. Disposable, problem free inoculation with smooth loop surface.
  • Meet semi-quantitative standard for handling samples
  • Surface treated for enhanced droplet adhesion
  • Non toxic
Specifications
  • Packed:
    50/sleeve
  • Capacity:
    -
  • Colour:
    Yellow
  • Description:
    Needles
  • Cat. No.:
    734-2139
  • Environmental certification:
    ISO 14001 (Certificate Registration No.CERT-IT000330)
  • Environmentally Preferable:
    Low Manufacturing Impact