Orders LinkContactUsLinkComponent
Replication systems, Nunc™
Replication systems, Nunc™
Replicator, 96-pin
Catalog # 732-2707
   Low Manufacturing Impact
Replication systems, Nunc™
Catalog # 732-2707
   Low Manufacturing Impact
Replication systems, Nunc™
Replication systems, Nunc™
Replicator, 96-pin
Price
4 180,00 €
Log in for availability
1 * 1 items
Product Details & Documents
Anodised aluminium replicators with stainless steel pins, PP copiers. Designed for the replication of DNA libraries, this system is ideal for transferring small volumes of liquid from plate to plate or to a membrane. Pins transfer 1,0 µl (±0,1 µl) of liquid.
Specifications
  • Description:
    Replicator, 96-pin
  • Seq:
    01
  • Cat. No.:
    732-2707
  • Environmental certification:
    NAR Certificate of Retirement
  • Environmentally Preferable:
    Low Manufacturing Impact