Replication systems, Nunc™
Replicator, 384-pin
Catalog # 732-2706
Supplier: Thermo Fisher Scientific
Replication systems, Nunc™
Catalog # 732-2706
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Anodised aluminium replicators with stainless steel pins, PP copiers. Designed for the replication of DNA libraries, this system is ideal for transferring small volumes of liquid from plate to plate or to a membrane. Pins transfer 1,0 µl (±0,1 µl) of liquid.
Specifications
- Description:Replicator, 384-pin
- Seq:02
- Cat. No.:732-2706
- Environmental certification:NAR Certificate of Retirement
- Environmentally Preferable:Low Manufacturing Impact