Orders LinkContactUsLinkComponent
MiniTrays, Nunclon™Δ
MiniTrays, Nunclon™Δ
MiniTray 60-well, conical well shape, Clear, 8 µl
Catalog # 734-2079
   Low Manufacturing Impact
MiniTrays, Nunclon™Δ
Catalog # 734-2079
   Low Manufacturing Impact
MiniTrays, Nunclon™Δ
MiniTrays, Nunclon™Δ
MiniTray 60-well, conical well shape, Clear, 8 µl
Price
522,00 €
Log in for availability
1 * 150 items
Product Details & Documents
PS, with lid, sterile.
External L×W: 84×59 mm
Specifications
  • Recommended working volume:
    8 µl
  • Colour:
    Clear
  • Description:
    MiniTray 60-well, conical well shape
  • Cat. No.:
    734-2079
  • Environmental certification:
    ISO 14001 (Certificate Registration No.CERT-IT000330)
  • Environmentally Preferable:
    Low Manufacturing Impact