Orders LinkContactUsLinkComponent
Syringes with valves
Syringes with valves
Syringe pre-fitted with Luer lock valve, PTFE tipped plunger, 50 ml, 50MR-VLLMA-GT
Catalog # SGEA009670
Syringes with valves
Catalog # SGEA009670
Syringes with valves
Syringes with valves
Syringe pre-fitted with Luer lock valve, PTFE tipped plunger, 50 ml, 50MR-VLLMA-GT
Price
471,00 €
Log in for availability
1 * 1 items
Product Details & Documents
These syringes are pre-fitted with valves. They are ideal for sample storage and transportation or to pressurise samples prior to injection.
Specifications
  • Type:
    50MR-VLLMA-GT
  • Volume:
    50 ml
  • Description:
    Syringe pre-fitted with Luer lock valve, PTFE tipped plunger
  • Seq:
    11