Orders LinkContactUsLinkComponent
Syringes with valves
Syringes with valves
Syringe pre-fitted with Luer lock valve, PTFE tipped plunger, 25 ml, 25MDR-VLLMA-GT
Catalog # SGEA009472
Syringes with valves
Catalog # SGEA009472
Syringes with valves
Syringes with valves
Syringe pre-fitted with Luer lock valve, PTFE tipped plunger, 25 ml, 25MDR-VLLMA-GT
Price
378,00 €
Log in for availability
1 * 1 items
Product Details & Documents
These syringes are pre-fitted with valves. They are ideal for sample storage and transportation or to pressurise samples prior to injection.
Specifications
  • Type:
    25MDR-VLLMA-GT
  • Volume:
    25 ml
  • Description:
    Syringe pre-fitted with Luer lock valve, PTFE tipped plunger
  • Seq:
    10