Syringes with valves
Syringe pre-fitted with Luer lock valve, PTFE tipped plunger, 10 ml, 10MDR-VLLMA-GT
Catalog # SGEA008970
Supplier: Trajan Scientific and Medical
Jump to:
Product Details & Documents
These syringes are pre-fitted with valves. They are ideal for sample storage and transportation or to pressurise samples prior to injection.
Specifications
- Type:10MDR-VLLMA-GT
- Volume:10 ml
- Description:Syringe pre-fitted with Luer lock valve, PTFE tipped plunger
- Seq:09
- Cat. No.:SGEA008970