Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
SPECTRA/POR 6 DIALYSIS TUBING 11.5 MM MW 1 * 10 m
Catalog # SERA44192.01
Supplier:
Serva
Expand image
SPECTRA/POR 6 DIALYSIS TUBING 11.5 MM MW 1 * 10 m
Catalog # SERA44192.01
Supplier:
Serva