Orders LinkContactUsLinkComponent
Gel drying frame kit
Gel drying frame kit
Gel drying frames (2), 100×100 mm, plus cellophane sheets
Catalog # SERA42523.01
Supplier:   Serva
Gel drying frame kit
Catalog # SERA42523.01
Supplier:  Serva
Gel drying frame kit
Gel drying frames (2), 100×100 mm, plus cellophane sheets
Price
148,00 €
Log in for availability
1 * 1 KIT
Product Details & Documents
Gel drying frame kit suitable for drying mini vertical slab gels up to 100×100 mm.
Delivery information: Available as frames only or as complete kit including two frames and 200 cellophane sheets.
Specifications
  • Description:
    Gel drying frames (2), 100×100 mm, plus cellophane sheets
  • Seq:
    01