Gel drying frame kit
Gel drying frames (2), 100×100 mm
Catalog # SERA42521.01
Supplier: Serva
Jump to:
Product Details & Documents
Gel drying frame kit suitable for drying mini vertical slab gels up to 100×100 mm.
Delivery information: Available as frames only or as complete kit including two frames and 200 cellophane sheets.
Specifications
- Description:Gel drying frames (2), 100×100 mm
- Seq:02