Holders for cryotubes, CryoCane™, Nalgene®
Colour code tabs, White
Catalog # 479-3270
Supplier: Thermo Fisher Scientific
Holders for cryotubes, CryoCane™, Nalgene®
Catalog # 479-3270
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Aluminium. Can be marked with colour code tabs.
Documents
Specifications
- Colour:White
- Description:Colour code tabs
- Seq:03
- Cat. No.:479-3270
- Environmental certification:NAR Certificate of Retirement
- Environmentally Preferable:Low Manufacturing Impact