Order Entry
Export
Orders LinkContactUsLinkComponent
Vacuum filtering flasks, Büchner, with side arm, Nalgene®

Vacuum filtering flasks, Büchner, with side arm, Nalgene®

1700 ml
Catalog # 215-8248
   Low Manufacturing Impact
Vacuum filtering flasks, Büchner, with side arm, Nalgene®
Catalog # 215-8248
   Low Manufacturing Impact
Vacuum filtering flasks, Büchner, with side arm, Nalgene®
Vacuum filtering flasks, Büchner, with side arm, Nalgene®

1700 ml

Price
103,00 €
Log in for availability
1 * 1 items
Product Details & Documents

PP, white.

  • Tubulation at 45° angle prevents tipping
  • Barbed adapter accepts 6,4 to 9,5 mm internal diameter tubing
  • Oval marking area
  • 215-8248 is graduated

Moulded in one piece to withstand full vacuum to 30-in. Hg.

Specifications
  • Capacity:
    1700 ml
  • Cat. No.:
    215-8248
  • Environmental certification:
    NAR Certificate of Retirement
  • Environmentally Preferable:
    Low Manufacturing Impact