Orders LinkContactUsLinkComponent
Sealing caps, Nalgene®
63 mm, 450/1000 ml
  525-2926
   Low Manufacturing Impact
Sealing caps, Nalgene®
  525-2926
   Low Manufacturing Impact
Sealing caps, Nalgene®
Sealing caps, Nalgene®
63 mm, 450/1000 ml
72,80 €
1 * 2 items
PP, autoclavable.
  • Silicone gasket
For use at recommended maximum speeds 10000×g without leakage, even when full.
  • For Ø ext.:
    63 mm
  • For tubes:
    450/1000 ml
  • Seq:
    04
  • Cat. No.:
    525-2926
  • Environmental certification:
    NAR Certificate of Retirement
  • Environmentally Preferable:
    Low Manufacturing Impact