Order Entry
Export
Orders LinkContactUsLinkComponent
Sealing caps, Nalgene®

Sealing caps, Nalgene®

58 mm, 250 ml
Catalog # 525-2925
   Low Manufacturing Impact
Sealing caps, Nalgene®
Catalog # 525-2925
   Low Manufacturing Impact
Sealing caps, Nalgene®
Sealing caps, Nalgene®

58 mm, 250 ml

Price
53,50 €
Log in for availability
1 * 2 items
Product Details & Documents

PP, autoclavable.

  • Silicone gasket

For use at recommended maximum speeds 10000×g without leakage, even when full.

Specifications
  • For Ø ext.:
    58 mm
  • For tubes:
    250 ml
  • Seq:
    03
  • Cat. No.:
    525-2925
  • Environmental certification:
    NAR Certificate of Retirement
  • Environmentally Preferable:
    Low Manufacturing Impact