Sealing caps, Nalgene®
20 mm, 28/30 ml
Catalog # 525-2923
Supplier: Thermo Fisher Scientific
Sealing caps, Nalgene®
Catalog # 525-2923
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
PP, autoclavable.
- Silicone gasket
For use at recommended maximum speeds 10000×g without leakage, even when full.
Specifications
- For Ø ext.:20 mm
- For tubes:28/30 ml
- Seq:01
- Cat. No.:525-2923
- Environmental certification:NAR Certificate of Retirement
- Environmentally Preferable:Low Manufacturing Impact