Sealing cap assemblies, Nalgene®
24 mm, 50 ml
Catalog # 525-2914
Supplier: Thermo Fisher Scientific
Sealing cap assemblies, Nalgene®
Catalog # 525-2914
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Tefzel™ FEP plug, PP screw closure, Viton™ fluoroelastomer O-ring.
- PP screw closure, ETFE plug, Viton™ fluoroelastomer O-ring
- Top fits securely for use at maximum speeds without leakage
- Autoclavable/biohazard/leakproof
Process tubes at their highest rated speed level (10000×g or greater) without leakage, even when full. Sealing cap assemblies include a Viton™ fluoroelastomer O-ring. Also recommended when spinning hazardous materials compatible with centrifuge tube material.
Specifications
- For Ø ext.:24 mm
- For tubes:50 ml
- Seq:02
- Cat. No.:525-2914
- Environmental certification:NAR Certificate of Retirement
- Environmentally Preferable:Low Manufacturing Impact



