Sealing caps, Nalgene®
63 mm
Catalog # NALG71-3132-0063
Supplier: Thermo Fisher Scientific
Sealing caps, Nalgene®
Catalog # NALG71-3132-0063
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
PP, autoclavable.
- Silicone gasket
For use at recommended maximum speeds 10000×g without leakage, even when full.
Specifications
- For Ø ext.:63 mm
- Seq:06
- Cat. No.:NALG71-3132-0063
- Environmental certification:NAR Certificate of Retirement
- Environmentally Preferable:Low Manufacturing Impact