Orders LinkContactUsLinkComponent
Sealing caps, Nalgene®
Sealing caps, Nalgene®
58 mm
Catalog # NALG71-3132-0058
   Low Manufacturing Impact
Sealing caps, Nalgene®
Catalog # NALG71-3132-0058
   Low Manufacturing Impact
Sealing caps, Nalgene®
Sealing caps, Nalgene®
58 mm
Price
60,30 €
Log in for availability
1 * 10 items
Product Details & Documents
PP, autoclavable.
  • Silicone gasket
For use at recommended maximum speeds 10000×g without leakage, even when full.
Specifications
  • For Ø ext.:
    58 mm
  • Seq:
    05
  • Cat. No.:
    NALG71-3132-0058
  • Environmental certification:
    NAR Certificate of Retirement
  • Environmentally Preferable:
    Low Manufacturing Impact