Stopcocks, for storage tanks, Nalgene®
19 mm NPT, male screw thread
Catalog # NALG6420-0750
Supplier: Thermo Fisher Scientific
Stopcocks, for storage tanks, Nalgene®
Catalog # NALG6420-0750
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
LDPE, white.
- Good chemical resistance
- Godd temperature resistance
Ensure safe and efficient liquid dispensing from tanks by using Nalgene™ spigots for storage tanks. Spigots are for use with Naglene™ tanks up to 100 gal. with factory installed threaded boss.
Specifications
- Thread:19 mm NPT, male screw thread
- Cat. No.:NALG6420-0750
- Environmental certification:NAR Certificate of Retirement
- Environmentally Preferable:Low Manufacturing Impact