Orders LinkContactUsLinkComponent
Stopcocks, for storage tanks, Nalgene®
Stopcocks, for storage tanks, Nalgene®
12,7 mm NPT, male thread
Catalog # NALG6420-0500
   Low Manufacturing Impact
Stopcocks, for storage tanks, Nalgene®
Catalog # NALG6420-0500
   Low Manufacturing Impact
Stopcocks, for storage tanks, Nalgene®
Stopcocks, for storage tanks, Nalgene®
12,7 mm NPT, male thread
Price
62,00 €
Log in for availability
1 * 1 items
Product Details & Documents
LDPE, white.
  • Good chemical resistance
  • Godd temperature resistance
Ensure safe and efficient liquid dispensing from tanks by using Nalgene™ spigots for storage tanks. Spigots are for use with Naglene™ tanks up to 100 gal. with factory installed threaded boss.
Specifications
  • Thread:
    12,7 mm NPT, male thread
  • Cat. No.:
    NALG6420-0500
  • Environmental certification:
    NAR Certificate of Retirement
  • Environmentally Preferable:
    Low Manufacturing Impact