Cryoware marker set, Nalgene®
Cryoware marker set, Black
Supplier: Thermo Fisher Scientific
Cryoware marker set, Nalgene®
Catalog # 479-0074
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
For permanent marking of cardboard, cloth labels and plastic.
- Ink will not fade at ultra-low temperatures
- Extra fine point
Set consists of 1 red, 1 green, 1 blue and 1 black marker.
Documents
Specifications
- Colour:Black
- Description:Cryoware marker set
- Cat. No.:479-0074
- Environmental certification:NAR Certificate of Retirement
- Environmentally Preferable:Low Manufacturing Impact