Orders LinkContactUsLinkComponent
Laboratory markers, Nalgene®
Laboratory markers, Nalgene®
Marker, thick tip, Black
Catalog # 479-0014
   Low Manufacturing Impact
Laboratory markers, Nalgene®
Catalog # 479-0014
   Low Manufacturing Impact
Laboratory markers, Nalgene®
Laboratory markers, Nalgene®
Marker, thick tip, Black
Price
27,20 €
Log in for availability
1 * 4 items
Product Details & Documents
Universal black pens for a wide variety of applications. Cap with handy clip.
  • Special fast drying ink
  • Highly chemical resistant and waterproof when writing on PolyPaper
  • Also suitable for marking glass and plastics
Specifications
  • Colour:
    Black
  • Description:
    Marker, thick tip
  • Cat. No.:
    479-0014
  • Environmental certification:
    NAR Certificate of Retirement
  • Environmentally Preferable:
    Low Manufacturing Impact