Laboratory markers, Nalgene®
Marker, fine tip, Black
Supplier: Thermo Fisher Scientific
Laboratory markers, Nalgene®
Catalog # 479-0013
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Universal black pens for a wide variety of applications. Cap with handy clip.
- Special fast drying ink
- Highly chemical resistant and waterproof when writing on PolyPaper
- Also suitable for marking glass and plastics
Specifications
- Colour:Black
- Description:Marker, fine tip
- Cat. No.:479-0013
- Environmental certification:NAR Certificate of Retirement
- Environmentally Preferable:Low Manufacturing Impact