Orders LinkContactUsLinkComponent
VWR®, Block Weights, Iron, Class M2
VWR®, Block Weights, Iron, Class M2
50 kg, 7500 ± mg
Catalog # 611-8390
Supplier:   VWR Collection
VWR®, Block Weights, Iron, Class M2
Catalog # 611-8390
Supplier:  VWR Collection
VWR®, Block Weights, Iron, Class M2
VWR®, Block Weights, Iron, Class M2
50 kg, 7500 ± mg
Price
576,00 €
Log in for availability
1 * 1 items
Product Details & Documents
Rectangular weights made from cast iron, permanently magnetised.
  • Indelible unique identification number guarantees traceability
  • Painted grey
  • Adjustment cavity sealed with a lead plug
 : Conforms to OIML, COFRAC accredited.
Specifications
  • Weight:
    50 kg
  • Seq:
    04
  • Tolerance:
    7500 ± mg