Order Entry
Export
Orders LinkContactUsLinkComponent
Pipette jars, with lid, Nalgene®

Pipette jars, with lid, Nalgene®

Pipette jar, HDPE, Hר: 514×165 mm
Catalog # 612-1803
   Low Manufacturing Impact
Pipette jars, with lid, Nalgene®
Catalog # 612-1803
   Low Manufacturing Impact
Pipette jars, with lid, Nalgene®
Pipette jars, with lid, Nalgene®

Pipette jar, HDPE, Hר: 514×165 mm

Price
259,00 €
Log in for availability
1 * 1 items
Product Details & Documents

Made of cross-linked high-density polyethylene, with PPCO base.

  • Tough, stiff, seamless jars with dust cover resist stress-cracking
  • Ideal for soaking glassware, washing pipets with corresponding Nalgene® rinsers and baskets
  • Flanged base for added stability

Please note: The formula shown on the jar's label is effective and lasts longer than standard solutions. Do not mix this solution in the pipet jar. Keep jar on a drainboard, in a sink or in a container large enough to retain jar contents in case of leakage.

Caution: Concentrated sulfuric acid will cause pipet jar to fail. When cleaning pipets with a sulfuric acid-dichromate compound, the acid concentration must be 60% or less.

Specifications
  • Description:
    Pipette jar, HDPE, Hר: 514×165 mm
  • Cat. No.:
    612-1803
  • Environmental certification:
    NAR Certificate of Retirement
  • Environmentally Preferable:
    Low Manufacturing Impact