VWR®, Block Weights, Iron, Class M2
5 kg, 750 ± mg
611-8383
Jump to:
Rectangular weights made from cast iron, permanently magnetised.
- Indelible unique identification number guarantees traceability
- Painted grey
- Adjustment cavity sealed with a lead plug
Certifications: Conforms to OIML, COFRAC accredited.
Documents
Specifications
- Weight:5 kg
- Seq:05
- Tolerance:750 ± mg