Orders LinkContactUsLinkComponent
VWR®, Calibration Weights, Class M2
VWR®, Calibration Weights, Class M2
5 kg, 750 ± mg
Catalog # 611-8381
Supplier:   VWR Collection
VWR®, Calibration Weights, Class M2
Catalog # 611-8381
Supplier:  VWR Collection
VWR®, Calibration Weights, Class M2
VWR®, Calibration Weights, Class M2
5 kg, 750 ± mg
Price
488,00 €
Log in for availability
1 * 1 items
Product Details & Documents
Test weights in either stainless steel or aluminium.
Delivery information: Supplied in a plastic box.
Specifications
  • Weight:
    5 kg
  • Seq:
    15
  • Tolerance:
    750 ± mg