VWR®, Calibration Weights, Class M2
50 g, 10 ± mg
Catalog # 611-8375
Supplier: VWR Collection
Jump to:
Product Details & Documents
Test weights in either stainless steel or aluminium.
: Supplied in a plastic box.
Documents
Specifications
- Weight:50 g
- Seq:09
- Tolerance:10 ± mg