VWR®, Calibration Weights, Class M2
20 g, 8 ± mg
Catalog # 611-8374
Supplier: VWR Collection
Jump to:
Product Details & Documents
Test weights in either stainless steel or aluminium.
Delivery information: Supplied in a plastic box.
Documents
Specifications
- Weight:20 g
- Seq:08
- Tolerance:8 ± mg