Orders LinkContactUsLinkComponent
VWR®, Calibration Weights, Class M2
1 g, 3 ± mg
Catalog # 611-8370
Supplier:   VWR Collection
VWR®, Calibration Weights, Class M2
Catalog # 611-8370
Supplier:  VWR Collection
VWR®, Calibration Weights, Class M2
VWR®, Calibration Weights, Class M2
1 g, 3 ± mg
Price
48,90 €
Log in for availability
1 * 1 items
Product Details & Documents
Test weights in either stainless steel or aluminium.
 : Supplied in a plastic box.
Specifications
  • Weight:
    1 g
  • Seq:
    04
  • Tolerance:
    3 ± mg