Order Entry
Export
Orders LinkContactUsLinkComponent
VWR®, Calibration Weights, Class M1

VWR®, Calibration Weights, Class M1

100 g, 5 ± mg
Catalog # 611-8287
Supplier:   VWR Collection
VWR®, Calibration Weights, Class M1
Catalog # 611-8287
Supplier:  VWR Collection
VWR®, Calibration Weights, Class M1
VWR®, Calibration Weights, Class M1

100 g, 5 ± mg

Price
61,80 €
Log in for availability
1 * 1 items
Product Details & Documents

Weights from 1 to 50 g are monobloc weights, with larger weights having an adjusting cavity.

Delivery information: All weights are supplied in plastic cases.

Specifications
  • Weight:
    100 g
  • Tolerance:
    5 ± mg